AMBH324ABA26 Ambeed, Inc.

Asenapine maleate

  • MDL number MFCD00900588
  • Cas Number : 85650-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/85650-56-2.html

Safety Information

MSDS https://www.ambeed.com/products/85650-56-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart