AMBH46A962F7 Ambeed, Inc.

AG 99

  • MDL number MFCD00133901
  • Cas Number : 122520-85-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/122520-85-8.html

Safety Information

MSDS https://www.ambeed.com/products/122520-85-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart