AMBH46A9630C Ambeed, Inc.

(R)-(+)-Etomoxir Sodium

  • MDL number MFCD07787411
  • Cas Number : 828934-41-4
  • Molecular Weight : 320.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/828934-41-4.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/828934-41-4.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart