AMBH46A96315 Ambeed, Inc.

BIBF0775

  • MDL number MFCD25976645
  • Cas Number : 334951-90-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/334951-90-5.html

Safety Information

MSDS https://www.ambeed.com/products/334951-90-5.html
Availability details for
Quantity
  • Availability
  • Add to Cart