AMBH46A963F1 Ambeed, Inc.

Spectinomycin dihydrochloride

  • MDL number MFCD00070403
  • Cas Number : 21736-83-4
  • Molecular Weight : 405.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/21736-83-4.html

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/21736-83-4.html
pictograms GHS07
hstatements H302-H315-H319-H332-H335
Availability details for
Quantity
  • Availability
  • Add to Cart