AMBH46A963FA Ambeed, Inc.

Tyrphostin AG 879

  • MDL number MFCD00236450
  • Cas Number : 148741-30-4
  • Molecular Weight : 316.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/148741-30-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/148741-30-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart