AMBH46A96404 Ambeed, Inc.

Anidulafungin

  • MDL number MFCD00917070
  • Cas Number : 166663-25-8
  • Molecular Weight : 1140.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/166663-25-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/166663-25-8.html
pictograms GHS07
hstatements H302-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart