AMBH46A96409 Ambeed, Inc.

Rolapitant hydrochloride hydrate

  • MDL number MFCD23105917
  • Cas Number : 914462-92-3
  • Molecular Weight : 554.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/914462-92-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/914462-92-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart