AMBH46A9640C Ambeed, Inc.

Upadacitinib

  • MDL number MFCD30502663
  • Cas Number : 1310726-60-3
  • Molecular Weight : 380.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/1310726-60-3.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1310726-60-3.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart