AMBH46A9645E Ambeed, Inc.

Z-Semaxanib

  • MDL number MFCD09763655
  • Cas Number : 194413-58-6
  • Molecular Weight : 238.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/194413-58-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/194413-58-6.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart