AMBH46A96468 Ambeed, Inc.

Bufalin

  • MDL number MFCD00056525
  • Cas Number : 465-21-4
  • Molecular Weight : 386.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/465-21-4.html

Safety Information

signalword Danger
pcodes P264-P301+P310
MSDS https://www.ambeed.com/products/465-21-4.html
pictograms GHS06
hstatements H300
Availability details for
Quantity
  • Availability
  • Add to Cart