AMBH46A9648E Ambeed, Inc.

XCT790

  • MDL number MFCD08277031
  • Cas Number : 725247-18-7
  • Molecular Weight : 596.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/725247-18-7.html

Safety Information

signalword Warning
pcodes P264-P270-P273-P301+P312-P330
MSDS https://www.ambeed.com/products/725247-18-7.html
pictograms GHS07
hstatements H302-H413
Availability details for
Quantity
  • Availability
  • Add to Cart