AMBH46A964C9 Ambeed, Inc.

G418 Disulfate

  • MDL number MFCD28168024
  • Cas Number : 108321-42-2
  • Molecular Weight : 692.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 620
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/108321-42-2.html

Safety Information

signalword Warning
pcodes P261-P280
MSDS https://www.ambeed.com/products/108321-42-2.html
pictograms GHS07
hstatements H317-H335
Availability details for
Quantity
  • Availability
  • Add to Cart