AMBH46A964CA Ambeed, Inc.

GSK 4716

  • MDL number MFCD00567155
  • Cas Number : 101574-65-6
  • Molecular Weight : 282.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/101574-65-6.html

Safety Information

signalword Danger
pcodes P273-P280-P501
MSDS https://www.ambeed.com/products/101574-65-6.html
pictograms GHS07,GHS09
hstatements H317-H410
Availability details for
Quantity
  • Availability
  • Add to Cart