AMBH46A964DA Ambeed, Inc.

CFTR(inh)-172

  • MDL number MFCD01164609
  • Cas Number : 307510-92-5
  • Molecular Weight : 409.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/307510-92-5.html

Safety Information

signalword Danger
pcodes P261-P273-P280-P305+P351+P338
MSDS https://www.ambeed.com/products/307510-92-5.html
pictograms GHS07,GHS09
hstatements H315-H317-H319-H335-H410
Availability details for
Quantity
  • Availability
  • Add to Cart