AMBH46A964DF Ambeed, Inc.

Leukadherin-1

  • MDL number MFCD01151276
  • Cas Number : 344897-95-6
  • Molecular Weight : 421.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/344897-95-6.html

Safety Information

signalword Danger
pcodes P273
MSDS https://www.ambeed.com/products/344897-95-6.html
pictograms GHS07,GHS09
hstatements H302-H410
Availability details for
Quantity
  • Availability
  • Add to Cart