AMBH46A964E4 Ambeed, Inc.

AG-494

  • MDL number MFCD00209863
  • Cas Number : 133550-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/133550-35-3.html

Safety Information

MSDS https://www.ambeed.com/products/133550-35-3.html
Availability details for
Quantity
  • Availability
  • Add to Cart