AMBH46A964E8 Ambeed, Inc.

Lycopene

  • MDL number MFCD00017350
  • Cas Number : 502-65-8
  • Molecular Weight : 536.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/502-65-8.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/502-65-8.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart