AMBH46A964EA Ambeed, Inc.

Ampicillin sodium

  • MDL number MFCD00064313
  • Cas Number : 69-52-3
  • Molecular Weight : 371.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/69-52-3.html

Safety Information

signalword Danger
pcodes P261-P280-P342+P311
MSDS https://www.ambeed.com/products/69-52-3.html
pictograms GHS08
hstatements H317-H334
Availability details for
Quantity
  • Availability
  • Add to Cart