AMBH46A964ED Ambeed, Inc.

Sec-O-glucosylhamaudol

  • MDL number MFCD11042255
  • Cas Number : 80681-44-3
  • Molecular Weight : 438.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/80681-44-3.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/80681-44-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart