AMBH46A96500 Ambeed, Inc.

Licochalcone A

  • MDL number MFCD01417903
  • Cas Number : 58749-22-7
  • Molecular Weight : 338.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/58749-22-7.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/58749-22-7.html
pictograms GHS07
hstatements H302-H312-H332
Availability details for
Quantity
  • Availability
  • Add to Cart