AMBH46A96501 Ambeed, Inc.

Methyl 2-(2-(2-chloro-5-((R)-2-hydroxy-3-(methylamino)propoxy)phenyl)-6-(3,5-dimethylisoxazol-4-yl)-5-methylpyrimidin-4-yl)-2,7-diazaspiro[3.5]nonane-7-carboxylate

  • MDL number MFCD31657380
  • Cas Number : 1628830-21-6
  • Molecular Weight : 585.0943

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Physical Form solid
Shipping Temperature Keep in dark place,Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/1628830-21-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/1628830-21-6.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart