AMBH46A96507 Ambeed, Inc.

Alantolactone

  • MDL number MFCD00046915
  • Cas Number : 546-43-0
  • Molecular Weight : 232.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/546-43-0.html

Safety Information

signalword Warning
pcodes P261-P280
MSDS https://www.ambeed.com/products/546-43-0.html
pictograms GHS07
hstatements H317
Availability details for
Quantity
  • Availability
  • Add to Cart