AMBH46A9655D Ambeed, Inc.

PD0325901

  • MDL number MFCD08435926
  • Cas Number : 391210-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/391210-10-9.html

Safety Information

MSDS https://www.ambeed.com/products/391210-10-9.html
Availability details for
Quantity
  • Availability
  • Add to Cart