AMBH46A96563 Ambeed, Inc.

SANT-1

  • MDL number MFCD01827306
  • Cas Number : 304909-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/304909-07-7.html

Safety Information

MSDS https://www.ambeed.com/products/304909-07-7.html
Availability details for
Quantity
  • Availability
  • Add to Cart