AMBH46A9656D Ambeed, Inc.

Arctiin

  • MDL number MFCD00870630
  • Cas Number : 20362-31-6
  • Molecular Weight : 534.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/20362-31-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/20362-31-6.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart