AMBH46A96578 Ambeed, Inc.

5-O-Methylvisammioside

  • MDL number MFCD10566635
  • Cas Number : 84272-85-5
  • Molecular Weight : 452.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/84272-85-5.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/84272-85-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart