AMBH46A9658A Ambeed, Inc.

QNZ46

  • MDL number MFCD22666391
  • Cas Number : 1237744-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/1237744-13-6.html

Safety Information

MSDS https://www.ambeed.com/products/1237744-13-6.html
Availability details for
Quantity
  • Availability
  • Add to Cart