AMBH46A965FF Ambeed, Inc.

Cefozopran hydrochloride

  • MDL number MFCD00944908
  • Cas Number : 113981-44-5
  • Molecular Weight : 551.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/113981-44-5.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/113981-44-5.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart