AMBH46A96612 Ambeed, Inc.

SB-239063

  • MDL number MFCD03792786
  • Cas Number : 193551-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin China
Product Link https://www.ambeed.com/products/193551-21-2.html

Safety Information

MSDS https://www.ambeed.com/products/193551-21-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart