AMBH46A96617 Ambeed, Inc.

UK-5099

  • MDL number N/A
  • Cas Number : 56396-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/56396-35-1.html

Safety Information

MSDS https://www.ambeed.com/products/56396-35-1.html
Availability details for
Quantity
  • Availability
  • Add to Cart