AMBH46A96618 Ambeed, Inc.

Vidarabine H2O

  • MDL number MFCD00150980
  • Cas Number : 24356-66-9
  • Molecular Weight : 285.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/24356-66-9.html

Safety Information

signalword Danger
pcodes P201-P202-P280-P308+P313-P405-P501
MSDS https://www.ambeed.com/products/24356-66-9.html
pictograms GHS08
hstatements H361
Availability details for
Quantity
  • Availability
  • Add to Cart