AMBH46A9662E Ambeed, Inc.

Penicillin G potassium

  • MDL number MFCD00036193
  • Cas Number : 113-98-4
  • Molecular Weight : 372.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/113-98-4.html

Safety Information

signalword Warning
pcodes P280
MSDS https://www.ambeed.com/products/113-98-4.html
pictograms GHS07
hstatements H317
Availability details for
Quantity
  • Availability
  • Add to Cart