AMBH46A96690 Ambeed, Inc.

SGC0946

  • MDL number MFCD26523136
  • Cas Number : 1561178-17-3
  • Molecular Weight : 618.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Keep in dark place,Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/1561178-17-3.html

Safety Information

signalword Danger
pcodes P301+P310
MSDS https://www.ambeed.com/products/1561178-17-3.html
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart