AMBH46A9669C Ambeed, Inc.

Clindamycin phosphate

  • MDL number MFCD11982855
  • Cas Number : 24729-96-2
  • Molecular Weight : 504.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 758
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/24729-96-2.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/24729-96-2.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart