AMBH46A966A0 Ambeed, Inc.

Pivmecillinam hydrochloride

  • MDL number MFCD01682071
  • Cas Number : 32887-03-9
  • Molecular Weight : 476.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/32887-03-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/32887-03-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart