AMBH46A966A5 Ambeed, Inc.

Ciliobrevin A

  • MDL number MFCD01911757
  • Cas Number : 302803-72-1
  • Molecular Weight : 358.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/302803-72-1.html

Safety Information

signalword Danger
pcodes P273-P501
MSDS https://www.ambeed.com/products/302803-72-1.html
pictograms GHS07,GHS09
hstatements H302-H410
Availability details for
Quantity
  • Availability
  • Add to Cart