AMBH46A966AA Ambeed, Inc.

Baccatin III

  • MDL number MFCD00153921
  • Cas Number : 27548-93-2
  • Molecular Weight : 586.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/27548-93-2.html

Safety Information

signalword Danger
pcodes P201-P261-P305+P351+P338-P308+P313
MSDS https://www.ambeed.com/products/27548-93-2.html
pictograms GHS07,GHS08
hstatements H315-H319-H335-H350
Availability details for
Quantity
  • Availability
  • Add to Cart