AMBH46A966B0 Ambeed, Inc.

1-(3-Ethyl-5-methoxybenzo[d]thiazol-2(3H)-ylidene)propan-2-one

  • MDL number N/A
  • Cas Number : 300801-52-9
  • Molecular Weight : 249.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Keep in dark place,Sealed in dry,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/300801-52-9.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/300801-52-9.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart