AMBH46A966BE Ambeed, Inc.

TM5441

  • MDL number MFCD31382187
  • Cas Number : 1190221-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.ambeed.com/products/1190221-43-2.html

Safety Information

MSDS https://www.ambeed.com/products/1190221-43-2.html
Availability details for
Quantity
  • Availability
  • Add to Cart