AMBH46A966C2 Ambeed, Inc.

Cefaclor monohydrate

  • MDL number MFCD00071999
  • Cas Number : 70356-03-5
  • Molecular Weight : 385.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Storage temp. Keep in dark place,Inert atmosphere,2-8
Country of Origin China
Product Link https://www.ambeed.com/products/70356-03-5.html

Safety Information

signalword Warning
pcodes P305+P351+P338
MSDS https://www.ambeed.com/products/70356-03-5.html
pictograms GHS07
hstatements H319
Availability details for
Quantity
  • Availability
  • Add to Cart