AMBH46A966CC Ambeed, Inc.

Tivantinib

  • MDL number MFCD11977597
  • Cas Number : 905854-02-6
  • Molecular Weight : 369.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,Store in freezer, under -20
Country of Origin China
Product Link https://www.ambeed.com/products/905854-02-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/905854-02-6.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart