AMBH47A98673 Ambeed, Inc.

5-Bromo-2-fluoro-3-methylbenzaldehyde

  • MDL number MFCD11110261
  • Cas Number : 903875-64-9
  • Molecular Weight : 217.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Inert atmosphere,2-8°C
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/903875-64-9.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/903875-64-9.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart