AMBH580641F2 Ambeed, Inc.

2-(Chloromethyl)cyclopropane-1-carboxylic acid

  • MDL number MFCD24506559
  • Cas Number : 1779806-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.ambeed.com/products/1779806-94-8.html

Safety Information

MSDS https://www.ambeed.com/products/1779806-94-8.html
Availability details for
Quantity
  • Availability
  • Add to Cart