AMBH580641FF Ambeed, Inc.

tert-Butyl 2-hydroxypropanoate

  • MDL number MFCD21641883
  • Cas Number : 59854-10-3
  • Molecular Weight : 146.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 2-hydroxypropanoate
Inchi Code 1S/C7H14O3/c1-5(8)6(9)10-7(2,3)4/h5,8H,1-4H3
Inchi Key IXXMVXXFAJGOQO-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Sealed in dry,Room Temperature
Physical Form liquid
Shipping Temperature Normal
Country of Origin CN
Product Link https://www.ambeed.com/products/59854-10-3.html

Safety Information

signalword Warning
pcodes P261-P301+P312-P302+P352-P304+P340-P305+P351+P338
MSDS https://www.ambeed.com/products/59854-10-3.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart