AMBH58064202 Ambeed, Inc.

(S)-1,1-Dimethylpyrrolidin-1-ium-2-carboxylate

  • MDL number MFCD09837899
  • Cas Number : 471-87-4
  • Molecular Weight : 143.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/471-87-4.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/471-87-4.html
pictograms GHS07
hstatements H302
Availability details for
Quantity
  • Availability
  • Add to Cart