AMBH5806420F Ambeed, Inc.

1,2,4-Thiadiazol-3-amine

  • MDL number MFCD12827990
  • Cas Number : 56531-89-6
  • Molecular Weight : 101.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Keep in dark place,Inert atmosphere,Room temperature
Country of Origin China
Product Link https://www.ambeed.com/products/56531-89-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/56531-89-6.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart