AMBH58064211 Ambeed, Inc.

4-Bromo-3-fluoro-2-hydroxybenzoic acid

  • MDL number MFCD23703631
  • Cas Number : 1429321-82-3
  • Molecular Weight : 235.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.ambeed.com/products/1429321-82-3.html

Safety Information

MSDS https://www.ambeed.com/products/1429321-82-3.html
Availability details for
Quantity
  • Availability
  • Add to Cart