AMBH58064215 Ambeed, Inc.

(trans,trans)-4-Ethyl-4'-(4-(trifluoromethoxy)phenyl)-1,1'-bi(cyclohexane)

  • MDL number MFCD09842837
  • Cas Number : 135734-59-7
  • Molecular Weight : 354.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Sealed in dry,Room Temperature
Country of Origin China
Product Link https://www.ambeed.com/products/135734-59-7.html

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS https://www.ambeed.com/products/135734-59-7.html
pictograms GHS07
hstatements H302-H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart