AMBH5806421C Ambeed, Inc.

3-Methyl-4-isopropylaniline hydrochloride

  • MDL number MFCD00156562
  • Cas Number : 4534-11-6
  • Molecular Weight : 185.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 223�t N/A mmHg
bp 247�t 760 mmHg
Iupac Name 4-isopropyl-3-methylaniline hydrochloride
Inchi Code 1S/C10H15N.ClH/c1-7(2)10-5-4-9(11)6-8(10)3;/h4-7H,11H2,1-3H3;1H
Inchi Key VPOAQDSAPBSOLW-UHFFFAOYSA-N

Description

Melting Point 223�t N/A mmHg
Purity 97%
Flash Point 106
Storage temp. Inert atmosphere,Room Temperature
Physical Form solid
Shipping Temperature Normal
Country of Origin CN
Boiling point 247�t 760 mmHg
Product Link https://www.ambeed.com/products/4534-11-6.html

Safety Information

signalword Warning
pcodes P280-P305+P351+P338
MSDS https://www.ambeed.com/products/4534-11-6.html
pictograms GHS07
hstatements H317-H319
Availability details for
Quantity
  • Availability
  • Add to Cart